Thermo Scientific Chemicals

7-Bromoisoquinoline N-oxide, 98%, Thermo Scientific Chemicals

Catalog number: H25899.06
5 g, Each
Thermo Scientific Chemicals

7-Bromoisoquinoline N-oxide, 98%, Thermo Scientific Chemicals

Catalog number: H25899.06
5 g, Each
Quantity
Catalog number: H25899.06
also known as H25899-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
223671-18-9
IUPAC Name
7-bromoisoquinolin-2-ium-2-olate
Molecular Formula
C9H6BrNO
InChI Key
QLRDQGRZLHHBPE-UHFFFAOYSA-N
SMILES
[O-][N+]1=CC2=CC(Br)=CC=C2C=C1
Form
Powder
Assay (HPLC)
≥97.5%
Water Content (Karl Fischer Titration)
<1.0%
Melting Point (clear melt)
178-188?C
Proton NMR
Conform to structure

Description

7-Bromoisoquinoline N-oxide is the reactant in the direct regioselective phosphonation of heteroaryl N-oxides with H-phosphonates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromoisoquinoline N-oxide is the reactant in the direct regioselective phosphonation of heteroaryl N-oxides with H-phosphonates

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text