2,7-Dibromo-9-fluorenone, 96%, Thermo Scientific Chemicals
2,7-Dibromo-9-fluorenone, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,7-Dibromo-9-fluorenone, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H25919.06
also known as H25919-06
Price (USD)/ Each
115.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
115.00
Add to cart
2,7-Dibromo-9-fluorenone, 96%, Thermo Scientific Chemicals
Catalog numberH25919.06
Price (USD)/ Each
115.00
-
Add to cart
Chemical Identifiers
CAS14348-75-5
IUPAC Name2,7-dibromo-9H-fluoren-9-one
Molecular FormulaC13H6Br2O
InChI KeyCWGRCRZFJOXQFV-UHFFFAOYSA-N
SMILESBrC1=CC=C2C3=CC=C(Br)C=C3C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (HPLC)≥95.0%
Melting Point (clear melt)191.0-206.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only