Thermo Scientific Chemicals

Dichloro(diethylamino)phosphine, 97%, Thermo Scientific Chemicals

Catalog number: H25934.06
5 g, Each
Thermo Scientific Chemicals

Dichloro(diethylamino)phosphine, 97%, Thermo Scientific Chemicals

Catalog number: H25934.06
5 g, Each
Quantity
Catalog number: H25934.06
also known as H25934-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1069-08-5
IUPAC Name
(dichlorophosphanyl)diethylamine
Molecular Formula
C4H10Cl2NP
InChI Key
BPEMCEULJQTJMI-UHFFFAOYSA-N
SMILES
CCN(CC)P(Cl)Cl
Appearance (Color)
Clear, colorless to pale yellow
Assay (unspecified)
>96.0% (by Phosphorous NMR)
Comment
May contain small amounts of visible sediment.
Proton NMR
Conforms to structure
Phosphorus-31 NMR
Conforms to structure

Description

Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.

Solubility
Reacts violently in water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text