2-Bromo-4-chloro-1-fluorobenzene, 97%, Thermo Scientific Chemicals
2-Bromo-4-chloro-1-fluorobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4-chloro-1-fluorobenzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H25935.14
also known as H25935-14
Price (USD)/ Each
257.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
257.00
Add to cart
2-Bromo-4-chloro-1-fluorobenzene, 97%, Thermo Scientific Chemicals
Catalog numberH25935.14
Price (USD)/ Each
257.00
-
Add to cart
Chemical Identifiers
CAS1996-30-1
IUPAC Name2-bromo-4-chloro-1-fluorobenzene
Molecular FormulaC6H3BrClF
InChI KeyYFFUYGSLQXVHMB-UHFFFAOYSA-N
SMILESFC1=C(Br)C=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5510-1.5560 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only