Thermo Scientific Chemicals

4-Cyano-2-fluoropyridine, 97%, Thermo Scientific Chemicals

Catalog number: H25938.03
1 g, Each
Thermo Scientific Chemicals

4-Cyano-2-fluoropyridine, 97%, Thermo Scientific Chemicals

Catalog number: H25938.03
1 g, Each
Quantity
Catalog number: H25938.03
also known as H25938-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3939-14-8
IUPAC Name
2-fluoropyridine-4-carbonitrile
Molecular Formula
C6H3FN2
InChI Key
DYQMGPSBUBTNJZ-UHFFFAOYSA-N
SMILES
FC1=CC(=CC=N1)C#N
Melting Point (clear melt)
28.0-38.0?C
Appearance (Color)
Colorless to white
Form
Fused solid or clear liquid as melt
Assay (GC)
≥96.0%
Water Content (Karl Fischer Titration)
≤1.5%

Description

4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride, nitrogen oxides, strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text