4-Cyano-2-fluoropyridine, 97%, Thermo Scientific Chemicals
4-Cyano-2-fluoropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyano-2-fluoropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number H25938.14
also known as H25938-14
Price (USD)/ Each
418.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
418.00
Add to cart
4-Cyano-2-fluoropyridine, 97%, Thermo Scientific Chemicals
Catalog numberH25938.14
Price (USD)/ Each
418.00
-
Add to cart
Chemical Identifiers
CAS3939-14-8
IUPAC Name2-fluoropyridine-4-carbonitrile
Molecular FormulaC6H3FN2
InChI KeyDYQMGPSBUBTNJZ-UHFFFAOYSA-N
SMILESFC1=CC(=CC=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)28.0-38.0?C
Appearance (Color)Colorless to white
FormFused solid or clear liquid as melt
Assay (GC)≥96.0%
Water Content (Karl Fischer Titration)≤1.5%
4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride, nitrogen oxides, strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
Bandarage, U. K.; Come, J. H.; Green, J. Convenient synthesis of N-(4-(2-aminopyridin-4-yl)thiazol-2-yl)-2-phenylacetamides. Tetrahedron Lett. 2006, 47 (46), 8079-8081.