5-Formyl-4-methylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals
5-Formyl-4-methylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Formyl-4-methylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H25947.06
also known as H25947-06
Price (USD)/ Each
288.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
288.00
Add to cart
5-Formyl-4-methylthiophene-2-boronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH25947.06
Price (USD)/ Each
288.00
-
Add to cart
Chemical Identifiers
CAS352530-25-7
IUPAC Name(5-formyl-4-methylthiophen-2-yl)boronic acid
Molecular FormulaC6H7BO3S
InChI KeyQEIXJCOHLUKRPO-UHFFFAOYSA-N
SMILESCC1=C(SC(=C1)B(O)O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale orange to pink
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
FormPowder
Melting Point (clear melt)119.5-125.5?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only