6-Amino-3-cyano-2-methylpyridine, 97%, Thermo Scientific Chemicals
6-Amino-3-cyano-2-methylpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Amino-3-cyano-2-methylpyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H26000.06
also known as H26000-06
Price (USD)/ Each
211.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
211.00
Add to cart
6-Amino-3-cyano-2-methylpyridine, 97%, Thermo Scientific Chemicals
Catalog numberH26000.06
Price (USD)/ Each
211.00
-
Add to cart
Chemical Identifiers
CAS183428-90-2
IUPAC Name6-amino-2-methylpyridine-3-carbonitrile
Molecular FormulaC7H7N3
InChI KeyOZAIJUZOQOBQDW-UHFFFAOYSA-N
SMILESCC1=C(C=CC(N)=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to pale brown
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)160.0-170.0?C
6-Amino-3-cyano-2-methylpyridine is a useful nitrile building block for proteomics research and synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-3-cyano-2-methylpyridine is a useful nitrile building block for proteomics research and synthesis.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
RUO – Research Use Only