Thermo Scientific Chemicals

tert-Butyl N-allylcarbamate, 97%, Thermo Scientific Chemicals

Catalog number: H26014.18
50 g, Each
Thermo Scientific Chemicals

tert-Butyl N-allylcarbamate, 97%, Thermo Scientific Chemicals

Catalog number: H26014.18
50 g, Each
Quantity
Catalog number: H26014.18
also known as H26014-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78888-18-3
IUPAC Name
tert-butyl N-(prop-2-en-1-yl)carbamate
Molecular Formula
C8H15NO2
InChI Key
AWARHXCROCWEAK-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCC=C
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)
≥96.0%

Description

tert-Butyl N-allylcarbamate is used in the synthesis of isoxazolidines. Synthesis of 5-substituted thiazolidin-2-ones is from the reaction of xanthates and tert-butyl N-allylcarbamates .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl N-allylcarbamate is used in the synthesis of isoxazolidines. Synthesis of 5-substituted thiazolidin-2-ones is from the reaction of xanthates and tert-butyl N-allylcarbamates .

Solubility
Insoluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text