Thermo Scientific Chemicals

2-Chloro-4-methoxyphenylacetonitrile, 95%, Thermo Scientific Chemicals

Catalog number: H26021.06
5 g, Each
Thermo Scientific Chemicals

2-Chloro-4-methoxyphenylacetonitrile, 95%, Thermo Scientific Chemicals

Catalog number: H26021.06
5 g, Each
Quantity
Catalog number: H26021.06
also known as H26021-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
170737-93-6
Appearance (Color)
White to cream to pale brown
Form
Powder
Assay (GC)
≥94.0%
Melting Point (clear melt)
39.5-45.5?C

Description

2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text