Thermo Scientific Chemicals

4-(2-Pyrrolidinyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog number: H26051.MD
250 mg, Each
Thermo Scientific Chemicals

4-(2-Pyrrolidinyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog number: H26051.MD
250 mg, Each
Quantity
Catalog number: H26051.MD
also known as H26051-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128562-25-4
IUPAC Name
2-oxo-3-(thiophen-2-yl)propanoate
Molecular Formula
C7H5O3S
InChI Key
DFEKJNVSDUKUIR-UHFFFAOYSA-M
SMILES
[O-]C(=O)C(=O)CC1=CC=CS1
Appearance (Color)
Clear yellow
Form
Liquid
Assay (GC)
≥95.0%
Refractive Index
1.5465-1.5525 @ 20?C

Description

4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text