Thermo Scientific Chemicals

3,3,3-Trifluoro-2,2-dimethylpropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26053.03
1 g, Each
Thermo Scientific Chemicals

3,3,3-Trifluoro-2,2-dimethylpropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26053.03
1 g, Each
Quantity
Catalog number: H26053.03
also known as H26053-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
889940-13-0
IUPAC Name
3,3,3-trifluoro-2,2-dimethylpropanoic acid
Molecular Formula
C5H7F3O2
InChI Key
VMFSJVUPIXOCFO-UHFFFAOYSA-N
SMILES
CC(C)(C(O)=O)C(F)(F)F
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Melting Point (clear melt)
65.0-74.0?C
Form
Crystals or powder or crystalline powder

Description

3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text