Chemical Identifiers
CAS144222-34-4
IUPAC NameN-[(1R,2R)-2-azaniumyl-1,2-diphenylethyl]-4-methylbenzene-1-sulfonamide
Molecular FormulaC21H23N2O2S
InChI KeyUOPFIWYXBIHPIP-NHCUHLMSSA-O
SMILESCC1=CC=C(C=C1)S(=O)(=O)N[C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
View more