Thermo Scientific Chemicals

4-Cyanophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26062.06
5 g, Each
Thermo Scientific Chemicals

4-Cyanophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26062.06
5 g, Each
Quantity
Catalog number: H26062.06
also known as H26062-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5462-71-5
IUPAC Name
2-(4-cyanophenyl)acetic acid
Molecular Formula
C9H7NO2
InChI Key
WEBXRQONNWEETE-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(C=C1)C#N
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
147.0-156.0?C

Description

4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Soluble in CHCl3, THF, dioxane.

Notes
Storage temperature is 2 - 4°C and Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text