Thermo Scientific Chemicals

1-Bromo-3-n-hexylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: H26063.06
5 g, Each
Thermo Scientific Chemicals

1-Bromo-3-n-hexylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: H26063.06
5 g, Each
Quantity
Catalog number: H26063.06
also known as H26063-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38409-59-5
IUPAC Name
1-bromo-3-hexylbenzene
Molecular Formula
C12H17Br
InChI Key
HZUVRCRPECDFAT-UHFFFAOYSA-N
SMILES
CCCCCCC1=CC(Br)=CC=C1
Identification (FTIR)
Conforms
Refractive Index
1.5190-1.5240 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.0%

Description

1-Bromo-3-n-hexylbenzene is used as intermediates in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-n-hexylbenzene is used as intermediates in pharmaceutical industry.

Solubility
Insoluble in water. Fully miscible with ethanol, acetone, toluene, methylene chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text