N,N-Dimethyl-1-dodecylamine N-oxide, 95%, Thermo Scientific Chemicals
N,N-Dimethyl-1-dodecylamine N-oxide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethyl-1-dodecylamine N-oxide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26065.06
also known as H26065-06
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
Add to cart
N,N-Dimethyl-1-dodecylamine N-oxide, 95%, Thermo Scientific Chemicals
Catalog numberH26065.06
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1643-20-5
IUPAC NameN,N-dimethyldodecanamine oxide
Molecular FormulaC14H31NO
InChI KeySYELZBGXAIXKHU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC[N+](C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
Water Content (Karl Fischer Titration)≤3.0%
Melting Point (clear melt)117.0-124.0?C
N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Schellmann, K.; Preisig, N.; Claesson, P.; Stubenrauch, C. Effects of protonation on foaming properties of dodecyldimethylamine oxide solutions: a pH-study. Soft Matter 2015, 11 (3), 561-571.
  2. Ghorbani, M.; Salehi, F.; Razavizadeh, O. Enhanced Hardness and Corrosion Resistance of Zn/SiO2 Films by Electrodeposition. J. Electrochem. Soc. 2015, 162 (9), D480-D485.