Thermo Scientific Chemicals

3-Pyrroline, tech. 85% (remainder pyrrolidine), Thermo Scientific Chemicals

Catalog number: H26072.06
5 g, Each
Thermo Scientific Chemicals

3-Pyrroline, tech. 85% (remainder pyrrolidine), Thermo Scientific Chemicals

Catalog number: H26072.06
5 g, Each
Quantity
Catalog number: H26072.06
also known as H26072-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-96-6
IUPAC Name
2,5-dihydro-1H-pyrrole
Molecular Formula
C4H7N
InChI Key
JVQIKJMSUIMUDI-UHFFFAOYSA-N
SMILES
C1NCC=C1
Water Content (Karl Fischer Titration)
≤2.0%
Refractive Index
1.4650-1.4730 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Assay (unspecified)
≥83.0% with pyrrolidine
Form
Liquid

Description

3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.

Solubility
Soluble in alcohol and ether. Miscible with water.

Notes
Air sensitive. Store at -20°C. Store away from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text