3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.
Solubility
Soluble in alcohol and ether. Miscible with water.
Notes
Air sensitive. Store at -20°C. Store away from oxidizing agents, air.
RUO – Research Use Only
Martin P. Green; Jeremy C. Prodger; Alexandra E. Sherlock and Christopher J. Hayes. A Convenient Method for 3-Pyrroline Synthesis. Org. Lett. 2001, 3 (21), 1-4.
Masahiro Sai and Seijiro Matsubara. Silver-Catalyzed Intramolecular Chloroamination of Allenes: Easy Access to Functionalized 3-Pyrroline and Pyrrole Derivatives. Org. Lett. 2011, 13 (17), 4676-4679.