Thermo Scientific Chemicals

2-Fluoro-6-methoxybenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: H26136.MD
250 mg, Each
Thermo Scientific Chemicals

2-Fluoro-6-methoxybenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: H26136.MD
250 mg, Each
Quantity
Catalog number: H26136.MD
also known as H26136-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
500912-17-4
IUPAC Name
2-(bromomethyl)-1-fluoro-3-methoxybenzene
Molecular Formula
C8H8BrFO
InChI Key
PVKFBCBTHHTDEX-UHFFFAOYSA-N
SMILES
COC1=C(CBr)C(F)=CC=C1
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear pale yellow to yellow to orange to pale brown
Refractive Index
1.5555-1.5595 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text