Thermo Scientific Chemicals

3,5-Difluoro-4-hydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H26139.MD
250 mg, Each
Thermo Scientific Chemicals

3,5-Difluoro-4-hydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H26139.MD
250 mg, Each
Quantity
Catalog number: H26139.MD
also known as H26139-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118276-06-5
IUPAC Name
3,5-difluoro-4-hydroxybenzaldehyde
Molecular Formula
C7H4F2O2
InChI Key
SKOYTQILPMNZQO-UHFFFAOYSA-N
SMILES
OC1=C(F)C=C(C=O)C=C1F
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
119.0-126.0°C

Description

3,5-Difluoro-4-hydroxybenzaldehyde is used in the preparation of (Z)-2,6- difluoro-((2-methyl-5-oxooxazol-4(5H)-ylidene)methyl)phenylacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-4-hydroxybenzaldehyde is used in the preparation of (Z)-2,6- difluoro-((2-methyl-5-oxooxazol-4(5H)-ylidene)methyl)phenylacetate.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text