4-Chloro-3-fluoroaniline, 99%, Thermo Scientific Chemicals
4-Chloro-3-fluoroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-3-fluoroaniline, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H26148.03
also known as H26148-03
Price (USD)/ Each
84.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
84.10
Add to cart
4-Chloro-3-fluoroaniline, 99%, Thermo Scientific Chemicals
Catalog numberH26148.03
Price (USD)/ Each
84.10
-
Add to cart
Chemical Identifiers
CAS367-22-6
IUPAC Name4-chloro-3-fluoroaniline
Molecular FormulaC6H5ClFN
InChI KeyACMJJQYSPUPMPN-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White to pale cream or grey
Melting Point (clear melt)58.0-62.0?C
FormCrystals or powder or crystalline powder or lumpy/fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only