Thermo Scientific Chemicals

4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26154.MD
250 mg, Each
Thermo Scientific Chemicals

4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26154.MD
250 mg, Each
Quantity
Catalog number: H26154.MD
also known as H26154-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
878572-17-9
IUPAC Name
4-fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
Molecular Formula
C8H3F4NO4
InChI Key
XCZRYPKVIJEHSU-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=C(F)C(=C1)[N+]([O-])=O)C(F)(F)F
Form
Powder
Assay (Silylated GC)
≥96.0%
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
118.0-128.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text