4-Amino-3-fluorobenzonitrile, 99%, Thermo Scientific Chemicals
4-Amino-3-fluorobenzonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-fluorobenzonitrile, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26170.03
also known as H26170-03
Price (USD)/ Each
188.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
188.00
Add to cart
4-Amino-3-fluorobenzonitrile, 99%, Thermo Scientific Chemicals
Catalog numberH26170.03
Price (USD)/ Each
188.00
-
Add to cart
Chemical Identifiers
CAS63069-50-1
IUPAC Name4-amino-3-fluorobenzonitrile
Molecular FormulaC7H5FN2
InChI KeyRLMBRRQWBTWGMB-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1F)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)85.0-91.0?C
Assay (GC)≥98.5%
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only