Thermo Scientific Chemicals

2-Bromobenzimidazole, 99%, Thermo Scientific Chemicals

Catalog number: H26183.03
1 g, Each
Thermo Scientific Chemicals

2-Bromobenzimidazole, 99%, Thermo Scientific Chemicals

Catalog number: H26183.03
1 g, Each
Quantity
Catalog number: H26183.03
also known as H26183-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54624-57-6
IUPAC Name
2-bromo-1H-1,3-benzodiazole
Molecular Formula
C7H5BrN2
InChI Key
PHPYXVIHDRDPDI-UHFFFAOYSA-N
SMILES
BrC1=NC2=CC=CC=C2N1
Proton NMR
Conforms to structure
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%

Description

Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text