Thermo Scientific Chemicals

3-Chloro-5-fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: H26195.03
1 g, Each
Thermo Scientific Chemicals

3-Chloro-5-fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: H26195.03
1 g, Each
Quantity
Catalog number: H26195.03
also known as H26195-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
202982-70-5
IUPAC Name
3-chloro-5-fluorophenol
Molecular Formula
C6H4ClFO
InChI Key
ZMFBYTAAMHWQHD-UHFFFAOYSA-N
SMILES
OC1=CC(F)=CC(Cl)=C1
Refractive Index
1.5345-1.5385 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text