Thermo Scientific Chemicals

2,6-Dimethyl-4-nitroanisole, 99%, Thermo Scientific Chemicals

Catalog number: H26204.06
5 g, Each
Thermo Scientific Chemicals

2,6-Dimethyl-4-nitroanisole, 99%, Thermo Scientific Chemicals

Catalog number: H26204.06
5 g, Each
Quantity
Catalog number: H26204.06
also known as H26204-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14804-39-8
IUPAC Name
2-methoxy-1,3-dimethyl-5-nitrobenzene
Molecular Formula
C9H11NO3
InChI Key
HSDNHFOJTRMGER-UHFFFAOYSA-N
SMILES
COC1=C(C)C=C(C=C1C)[N+]([O-])=O
Form
Crystals or powder or crystalline powder
Comment
Yellow
Melting Point (clear melt)
87.0-93.0?C
Assay (GC)
≥98.5%
Water Content (Karl Fischer Titration)
≤1.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text