Thermo Scientific Chemicals

4-Bromo-2,5-difluorophenol, 99%, Thermo Scientific Chemicals

Catalog number: H26212.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-2,5-difluorophenol, 99%, Thermo Scientific Chemicals

Catalog number: H26212.06
5 g, Each
Quantity
Catalog number: H26212.06
also known as H26212-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
486424-36-6
IUPAC Name
4-bromo-2,5-difluorophenol
Molecular Formula
C6H3BrF2O
InChI Key
BYZMZJIWCQTYSR-UHFFFAOYSA-N
SMILES
OC1=CC(F)=C(Br)C=C1F
Appearance (Color)
White to cream or pale purple
Form
Crystals or powder or crystalline powder or flakes or lumpy/fused solid or chunks
Melting Point (clear melt)
48.5-54.5°C
Assay (GC)
≥98.5%

Description

4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.

Solubility
Soluble in water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text