3-Bromo-2,6-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals
3-Bromo-2,6-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-2,6-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H26213.14
also known as H26213-14
Price (USD)/ Each
575.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
575.00
Add to cart
3-Bromo-2,6-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberH26213.14
Price (USD)/ Each
575.00
-
Add to cart
Chemical Identifiers
CAS398456-82-1
IUPAC Name3-bromo-2,6-difluorobenzaldehyde
Molecular FormulaC7H3BrF2O
InChI KeyOXBHKEYDKAWFLS-UHFFFAOYSA-N
SMILESFC1=C(C=O)C(F)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Free acid (titration)≤1.5%
Melting Point (clear melt)52.0-58.0?C
3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
RUO – Research Use Only