Thermo Scientific Chemicals

2-Methylheptane, 99%, Thermo Scientific Chemicals

Catalog number: H26220.14
25 g, Each
Thermo Scientific Chemicals

2-Methylheptane, 99%, Thermo Scientific Chemicals

Catalog number: H26220.14
25 g, Each
Quantity
Catalog number: H26220.14
also known as H26220-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
592-27-8
IUPAC Name
2-methylheptane
Molecular Formula
C8H18
InChI Key
JVSWJIKNEAIKJW-UHFFFAOYSA-N
SMILES
CCCCCC(C)C
Form
Liquid
Refractive Index
1.3930-1.3960 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text