Thermo Scientific Chemicals

tert-Butyl hydrogen malonate, 96%, Thermo Scientific Chemicals

Catalog number: H26223.14
25 g, Each
Thermo Scientific Chemicals

tert-Butyl hydrogen malonate, 96%, Thermo Scientific Chemicals

Catalog number: H26223.14
25 g, Each
Quantity
Catalog number: H26223.14
also known as H26223-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40052-13-9
IUPAC Name
3-(tert-butoxy)-3-oxopropanoic acid
Molecular Formula
C7H12O4
InChI Key
NGGGZUAEOKRHMA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)CC(O)=O
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Assay (Silylated GC)
≥95.0%
Identification (FTIR)
Conforms

Description

tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

Solubility
Soluble in ethanol.

Notes
Store in a cool, dry condition in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text