4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water 9.59 g/ l.
Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Brian John. Johnson; Therese A. N-Acylamino acid esters of 4-(methylthio)phenol. J. Org. Chem. 1970, 35 (1), 255-257.
- Brian J. Johnson; Elwood G. Trask. 4-(Methylthio)phenyl and 4-(methylsulfonyl)phenyl esters in the preparation of peptides and polypeptides. II. Synthesis of the protected heptapeptide (A82-A88) of bovine chymotrypsinogen A. J. Org. Chem. 1968, 33 (12), 4521-4524.