Vinylcyclopentane, 99%, Thermo Scientific Chemicals
Vinylcyclopentane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinylcyclopentane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26230.03
also known as H26230-03
Price (USD)/ Each
71.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
71.20
Add to cart
Vinylcyclopentane, 99%, Thermo Scientific Chemicals
Catalog numberH26230.03
Price (USD)/ Each
71.20
-
Add to cart
Chemical Identifiers
CAS3742-34-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Solution Test(10% v/v in diethyl ether) : Clear
Refractive Index1.4355-1.4385 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only