1-Bromo-4-n-propylbenzene, 99%, Thermo Scientific Chemicals
1-Bromo-4-n-propylbenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-4-n-propylbenzene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H26236.06
also known as H26236-06
Price (USD)/ Each
38.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.00
Add to cart
1-Bromo-4-n-propylbenzene, 99%, Thermo Scientific Chemicals
Catalog numberH26236.06
Price (USD)/ Each
38.00
-
Add to cart
Chemical Identifiers
CAS588-93-2
IUPAC Name1-bromo-4-propylbenzene
Molecular FormulaC9H11Br
InChI KeyNUPWGLKBGVNSJX-UHFFFAOYSA-N
SMILESCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5345-1.5375 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only