4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.
Solubility
Soluble in organic solvents.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Jun, X.; Ahmed, E.; Xiao, B.; Lu, Q.; Wang, Y.; Yu, C.; Fu, Y. Pd-Catalyzed Regioselective Activation of gem-Difluorinated Cyclopropanes: A Highly Efficient Approach to 2-Fluorinated Allylic Scaffolds. Angew. Chem. 2015, 127 (28), 8349-8353.
Belfrage, A. K.; Gising, J.; Svensson, F.; Akerblom, E.; Skold, C.; Sandstrom, A. Efficient and Selective Palladium-Catalysed C-3 Urea Couplings to 3,5-Dichloro-2(1H)-pyrazinones. Eur. J. Org. Chem. 2015, 2015 (5), 978-986.