Thermo Scientific Chemicals

1-Dodecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H26259.09
10 g, Each
Thermo Scientific Chemicals

1-Dodecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H26259.09
10 g, Each
Quantity
Catalog number: H26259.09
also known as H26259-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5137-70-2
IUPAC Name
dodecylphosphonic acid
Molecular Formula
C12H27O3P
InChI Key
SVMUEEINWGBIPD-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCP(O)(O)=O
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥94% to ≤106.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale grey

Description

1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text