Thermo Scientific Chemicals

4-Fluorophenylacetylene, 99%, Thermo Scientific Chemicals

Catalog number: H26269.03
1 g, Each
Thermo Scientific Chemicals

4-Fluorophenylacetylene, 99%, Thermo Scientific Chemicals

Catalog number: H26269.03
1 g, Each
Quantity
Catalog number: H26269.03
also known as H26269-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
766-98-3
IUPAC Name
1-ethynyl-4-fluorobenzene
Molecular Formula
C8H5F
InChI Key
QXSWHQGIEKUBAS-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)C#C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5160-1.5190 @ 20?C

Description

Intermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.

Solubility
Soluble in chloroform, diethyl ether, acetone, dichloromethane, hexane and ethanol. Highly soluble in toluene. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heavy metal powders.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text