Intermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Intermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.
Solubility
Soluble in chloroform, diethyl ether, acetone, dichloromethane, hexane and ethanol. Highly soluble in toluene. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heavy metal powders.
RUO – Research Use Only
A. S. Peregudov,; V. F. Ivanov,; D. N. Kravtsov,; É. I. Fedin. Synthesis 9F-{1H} NMR spectra of 4-fluorophenylacetylene and its organometallic derivatives. Organic Chemistry. 1984, 33 (4), 783-787.
Marko Strukelj,; Martino Paventi,; Allan S. Hay. Novel poly(aryl ethers) from bis(4-fluorophenyl)acetylene. Macromolecules. 1993, 22 (7),1777-1778.