Intermediate in the preparation of chelating agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Intermediate in the preparation of chelating agents.
Solubility
Soluble in acetone, acetonitrile, ethyl Acetate, chloroforme, cyclohexane. Insoluble in water.
Notes
Hygroscopic. Store in cool place.
RUO – Research Use Only
General References:
- Cong Li,; Wing-Tak Wong. A Simple, Regioselective Synthesis of 1,4-Bis(tert-butoxycarbonylmethyl)- tetraazacyclododecane. J. Org. Chem. 2003, 68(7) ,2956-2959.
- Cong Li,; Wing-Tak Wong. A selective one-step synthesis of tris N-alkylated cyclens. Tetrahedron. 2004, 60 (26), 5595-5601.