Thermo Scientific Chemicals

4-Bromobenzyl mercaptan, 98%, Thermo Scientific Chemicals

Catalog number: H26299.MD
250 mg, Each
Thermo Scientific Chemicals

4-Bromobenzyl mercaptan, 98%, Thermo Scientific Chemicals

Catalog number: H26299.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H26299.MD
also known as H26299-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19552-10-4
IUPAC Name
(4-bromophenyl)methanethiol
Molecular Formula
C7H7BrS
InChI Key
CUCKXDPCCYHFMQ-UHFFFAOYSA-N
SMILES
SCC1=CC=C(Br)C=C1
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥97.5%
Form
Fused low melting solid (Clear to turbid liquid as melt)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text