4-Ethylphenylacetylene, 99%, Thermo Scientific Chemicals
4-Ethylphenylacetylene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethylphenylacetylene, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H26313.06
also known as H26313-06
Price (USD)/ Each
99.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
99.20
Add to cart
4-Ethylphenylacetylene, 99%, Thermo Scientific Chemicals
Catalog numberH26313.06
Price (USD)/ Each
99.20
-
Add to cart
Chemical Identifiers
CAS40307-11-7
IUPAC Name1-ethyl-4-ethynylbenzene
Molecular FormulaC10H10
InChI KeyZNTJVJSUNSUMPP-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5405-1.5435 @ 20°C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only