Diethyl (2,4-dichlorophenyl)malonate, 95%, Thermo Scientific Chemicals
Diethyl (2,4-dichlorophenyl)malonate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl (2,4-dichlorophenyl)malonate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Catalog number H26330.ME
also known as H26330-ME
Price (USD)/ Each
96.80
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
96.80
Add to cart
Diethyl (2,4-dichlorophenyl)malonate, 95%, Thermo Scientific Chemicals
Catalog numberH26330.ME
Price (USD)/ Each
96.80
-
Add to cart
Chemical Identifiers
CAS111544-93-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5095-1.5155 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only