Thermo Scientific Chemicals

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate, 97+%, Thermo Scientific Chemicals

Catalog number: H26333.06
5 g, Each
Thermo Scientific Chemicals

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate, 97+%, Thermo Scientific Chemicals

Catalog number: H26333.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H26333.06
also known as H26333-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3945-69-5
IUPAC Name
4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium chloride
Molecular Formula
C10H17ClN4O3
InChI Key
BMTZEAOGFDXDAD-UHFFFAOYSA-M
SMILES
[Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1
Form
Powder
Assay (Titration ex Chloride)
≥97.0 to ≤103.0% (Dry wt. basis)
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤14%
Appearance (Color)
White

Description

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate is used as a pharmaceutical intermediate. It is used as a reagent for activating carboxylic acids in solution and solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate is used as a pharmaceutical intermediate. It is used as a reagent for activating carboxylic acids in solution and solid phase peptide synthesis.

Solubility
Soluble in water, Methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text