Thermo Scientific Chemicals

1-Phenylethyl mercaptan, 98%, Thermo Scientific Chemicals

Catalog number: H26345.03
1 g, Each
Thermo Scientific Chemicals

1-Phenylethyl mercaptan, 98%, Thermo Scientific Chemicals

Catalog number: H26345.03
1 g, Each
Quantity
Catalog number: H26345.03
also known as H26345-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6263-65-6
IUPAC Name
1-phenylethane-1-thiol
Molecular Formula
C8H10S
InChI Key
QZZBJCFNHPYNKO-UHFFFAOYNA-N
SMILES
CC(S)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.5560-1.5600 @ 20?C
Form
Liquid

Description

The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

Solubility
Insoluble in water. Solubility in ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text