Thermo Scientific Chemicals

4'-Chloro-3'-fluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H26373.03
1 g, Each
Thermo Scientific Chemicals

4'-Chloro-3'-fluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H26373.03
1 g, Each
Quantity
Catalog number: H26373.03
also known as H26373-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151945-84-5
IUPAC Name
1-(4-chloro-3-fluorophenyl)ethan-1-one
Molecular Formula
C8H6ClFO
InChI Key
ATZHNDSADJDUPJ-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(Cl)C(F)=C1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
36.0-46.0?C

Description

4'-Chloro-3'-fluoroacetophenone is a halogenated benzene derivative used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Chloro-3′-fluoroacetophenone is a halogenated benzene derivative used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text