Thermo Scientific Chemicals

5,5'-Dimethyl-2,2'-bipyridine, 98%, Thermo Scientific Chemicals

Catalog number: H26378.06
5 g, Each
Thermo Scientific Chemicals

5,5'-Dimethyl-2,2'-bipyridine, 98%, Thermo Scientific Chemicals

Catalog number: H26378.06
5 g, Each
Quantity
Catalog number: H26378.06
also known as H26378-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1762-34-1
IUPAC Name
5,5'-dimethyl-2,2'-bipyridine
Molecular Formula
C12H12N2
InChI Key
PTRATZCAGVBFIQ-UHFFFAOYSA-N
SMILES
CC1=CC=C(N=C1)C1=CC=C(C)C=N1
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
113.0-119.0?C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

5,5'-Dimethyl-2,2'-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5’vinyl-2,2’-bipyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,5′-Dimethyl-2,2′-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5’vinyl-2,2’-bipyridine.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text