2-Fluoro-6-methoxyphenol, 97%, Thermo Scientific Chemicals
2-Fluoro-6-methoxyphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-6-methoxyphenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H26471.06
also known as H26471-06
Price (USD)/ Each
248.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
248.00
Add to cart
2-Fluoro-6-methoxyphenol, 97%, Thermo Scientific Chemicals
Catalog numberH26471.06
Price (USD)/ Each
248.00
-
Add to cart
Chemical Identifiers
CAS73943-41-6
IUPAC Name2-fluoro-6-methoxyphenol
Molecular FormulaC7H7FO2
InChI KeyYZNHPLVFLRSVHY-UHFFFAOYSA-N
SMILESCOC1=C(O)C(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5160-1.5210 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only