Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.
Solubility
Slightly soluble water. Soluble in chloroform.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
RUO – Research Use Only
N H Lin et. al. Structure-activity studies on 2-methyl-3-(2(S)-pyrrolidinylmethoxy) pyridine (ABT-089): an orally bioavailable 3-pyridyl ether nicotinic acetylcholine receptor ligand with cognition-enhancing properties. Journal of Medicinal Chemistry. 1997, 40 (3), 385-390 .