Powder or lumpy powder or crystals or crystalline powder and/or chunks
Assay (GC)
≥95.0%
Melting Point (clear melt)
81-90°C
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Figuras
Documentos y descargas
Certificados
Buscar por número de lote o por número de lote parcial