2-Chloro-4-methylquinoline, 99%, Thermo Scientific Chemicals
2-Chloro-4-methylquinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-4-methylquinoline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26504.06
also known as H26504-06
Price (USD)/ Each
55.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
55.00
Add to cart
2-Chloro-4-methylquinoline, 99%, Thermo Scientific Chemicals
Catalog numberH26504.06
Price (USD)/ Each
55.00
-
Add to cart
Chemical Identifiers
CAS634-47-9
IUPAC Name2-chloro-4-methylquinoline
Molecular FormulaC10H8ClN
InChI KeyPFEIMKNQOIFKSW-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)53.0-59.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only