Thermo Scientific Chemicals

2-Phenylthioacetamide, 97%, Thermo Scientific Chemicals

Catalog number: H26514.03
1 g, Each
Thermo Scientific Chemicals

2-Phenylthioacetamide, 97%, Thermo Scientific Chemicals

Catalog number: H26514.03
1 g, Each
Quantity
Catalog number: H26514.03
also known as H26514-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
645-54-5
IUPAC Name
2-phenylethanethioamide
Molecular Formula
C8H9NS
InChI Key
CJXBHFANXQMZBF-UHFFFAOYSA-N
SMILES
NC(=S)CC1=CC=CC=C1
Appearance (Color)
White to cream
Assay (HPLC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
93.5-99.5?C

Description

2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.

Solubility
Slightly Soluble in water (1.8 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text